Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 9 beta 1 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 104099 |
| Molecular Weight (g/mol) | 114 kDa (Integrin alpha9) & 86.5 kDa (Integrin beta1) |
| Name | Integrin alpha 9 beta 1 |
| Quantity | 50μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 9 beta 1 protein, (N-terminus) Mouse Integrin alpha 9 (Tyr31-Val979) Accession &Num; CAC69080 His-Pro GGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.