Learn More
R&D Systems™ Recombinant Mouse Integrin alpha V beta 3 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 115 kDa (Integrin alphaV) & 84.5 kDa (Integrin beta3) |
| Name | Integrin alpha V beta 3 |
| Quantity | 50μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 3 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 3 (Glu26-Asp717) Accession &Num; O54890 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE with silver staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.