Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha V beta 5 Protein

Catalog No. 7706AV050
Change view
Click to view available options
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
7706AV050 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 7706AV050 Supplier R&D Systems™ Supplier No. 7706AV050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Integrin alpha V beta 5 Protein is derived from CHO. The Recombinant Mouse Integrin alpha V beta 5 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 115 kDa ( alphaV subunit) & 84.8 kDa (beta5 subunit)
Name Integrin alpha V beta 5
Quantity 50μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 5 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 HP GS Linker Acidic Tail HHHHHH Mouse Integrin beta 5 (Gly24-Asn719) Accession &Num; NP_001139356 HP GS Linker Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.