Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant mouse interleukin-31 | >85.0% | 2 UG
SDP

Catalog No. 5000292892
Encompass_Preferred

Recombinant mouse interleukin-31 produced in HEK293 cells and supplied as a filtered PBS solution for research applications. The protein is provided as a solution (2 μg vial) with a C-terminal His tag and is characterized by predicted mass and receptor-binding activity suitable for in vitro assays.

  • Expressed in HEK293 mammalian cells for native folding and glycosylation.
  • C-terminal His tag facilitates purification and detection.
  • Predicted molecular mass approximately 17.1 kDa.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Demonstrated receptor binding activity (ED50 reported in functional assay).
  • Supplied in PBS, pH 7.4; store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-92892 Supplier Medchemexpress LLC Supplier No. HYP748042UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-31 produced in HEK293 cells and supplied as a filtered PBS solution for research applications. The protein is provided as a solution (2 μg vial) with a C-terminal His tag and is characterized by predicted mass and receptor-binding activity suitable for in vitro assays.

  • Expressed in HEK293 mammalian cells for native folding and glycosylation.
  • C-terminal His tag facilitates purification and detection.
  • Predicted molecular mass approximately 17.1 kDa.
  • Purity greater than 85% as determined by reducing SDS-PAGE.
  • Demonstrated receptor binding activity (ED50 reported in functional assay).
  • Supplied in PBS, pH 7.4; store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.