Learn More
Medchemexpress LLC Recombinant mouse interleukin-31 (IL-31) | >85.0% | 50 UG
SDP
Catalog No. 5000291185
Encompass_Preferred
Recombinant mouse interleukin-31 (IL-31) produced in HEK293 cells with a C-terminal His tag, supplied as a 0.22 μm filtered solution in PBS (pH 7.4). The protein is designed for biochemical and cell-based assays, including receptor binding and signaling studies; manufacturer provides activity, purity, and storage data.
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Supplied in PBS (pH 7.4), 0.22 μm filtered.
  • >85.0% purity by reducing SDS-PAGE.
  • Approximate molecular weight 17.1 kDa (reducing SDS-PAGE).
  • Bioactivity: ED50 ≈ 2.916 μg/mL in receptor-binding ELISA.
  • Store long-term at -80°C; avoid repeated freeze-thaw.
Catalog No. 50-002-91185 Supplier Medchemexpress LLC Supplier No. HYP7480450UG
Recombinant mouse interleukin-31 (IL-31) produced in HEK293 cells with a C-terminal His tag, supplied as a 0.22 μm filtered solution in PBS (pH 7.4). The protein is designed for biochemical and cell-based assays, including receptor binding and signaling studies; manufacturer provides activity, purity, and storage data.
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Supplied in PBS (pH 7.4), 0.22 μm filtered.
  • >85.0% purity by reducing SDS-PAGE.
  • Approximate molecular weight 17.1 kDa (reducing SDS-PAGE).
  • Bioactivity: ED50 ≈ 2.916 μg/mL in receptor-binding ELISA.
  • Store long-term at -80°C; avoid repeated freeze-thaw.