Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Klotho beta Protein

Catalog No. 2619KB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2619KB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2619KB050 Supplier R&D Systems™ Supplier No. 2619KB050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human FGF RIIIc. The ED50 for this effect is 0.2-1 μg/mL in the presence of Recombinant Human FGF-21 (Catalog # 2539-FG ) and Heparin.

Specifications

Accession Number NP_112457
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS, Glycerol and EDTA.
Gene ID (Entrez) 152831
Molecular Weight (g/mol) M.W. (Observed): 140 kDa, reducing conditions; M.W. (theoretical): 110.2 kDa
Name Klotho beta
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Klotho beta protein Phe53-Leu995, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Klb
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Klotho beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.