Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Lipocalin-13 Protein

Catalog No. 8534LC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-34L-C050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-34L-C050 Supplier R&D Systems™ Supplier No. 8534LC050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Lipocalin-13 Protein is derived from E. coli. The Recombinant Mouse Lipocalin-13 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8K1H9
For Use With (Application) Bioactivity
Gene ID (Entrez) 227627
Molecular Weight (g/mol) Observed M.W.: 18kDa, reducing conditions, Theoretical M.W.: 18kDa
Quantity 50 μg
Source E. coli-derived mouse Lipocalin-13 protein Ala19-Asp176
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name Lipocalin-13
Gene Symbol Obp2a
Biological Activity Measured by its ability to bind all-trans-retinoic acid. The concentration of all-trans-retinoic acid required to quench 50% of Trp fluorescence in Recombinant Mouse Lipocalin-13 is approximately 3-18μM.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Activity Measured by its ability to bind all-trans-retinoic acid. The concentration of all-trans-retinoic acid required to quench 50% of Trp fluorescence in Recombinant Mouse Lipocalin-13 is approximately 20-120 μM.
Expression System E. coli-derived Ala19-Asp176
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.