Learn More
R&D Systems™ Recombinant Mouse LRP-6 N-Terminal Fragment Fc Protein
Description
Specifications
Specifications
| Accession Number | NP_032540 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 4040 |
| Molecular Weight (g/mol) | 96 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse LRP-6 protein, (N-terminus) Mouse LRP-6 (Ala20-Ile628) Accession &Num; NP_032540 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.