Learn More
Invivogen Recombinant Mouse Anti-Mouse IL-13 mAb, Neutralizes IL-13, 100% Murine Sequence, 1 mg
SDP
Supplier:  Invivogen MIL13MAB91
Encompass
Recombinant Mouse Anti-Mouse IL-13 mAb is engineered for potent neutralization of murine IL-13, enhancing inflammatory response studies. Produced in CHO cells, it offers lot-to-lot reproducibility, low immunogenicity, and reliability for in vivo applications. Available in InvivoFit™ grade, it ensures low endotoxin levels. This antibody's all-murine sequence minimizes immunogenicity, making it an ideal choice for research requiring precise cytokine targeting.
Catalog No. NC2243215