Learn More
R&D Systems™ Recombinant Mouse Neuroplastin Protein
Description
Specifications
Specifications
| Concentration | >95% by SDS-PAGE w/silver stain |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Neuroplastin protein Gln29-Pro338,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.