Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse OLT-2/TARM1 Fc Chimera Protein

Catalog No. 10000TA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-000-TA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-000-TA050 Supplier R&D Systems™ Supplier No. 10000TA050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Recombinant Mouse OLT-2/TARM1 Fc Chimera Protein is derived from NS0

  • TARM-1 (T cell interacting, activating receptor on myeloid cells-1), also name OLT-2 (OSCAR-like Transcript 2), is a type 1 membrane protein expressed by neutrophils, inflammatory monocytes, macrophages, and dendritic cells.
  • Mature mouse TARM-1 contains a 242 amino acid (aa) long extracellular domain with two Ig-like C2 domains, a 21 aa long transmembrane domain and a 9 aa long cytoplasmic domain.
  • The extracellular domain of mouse TARM-1 shares 81.6% and 46.5% aa identity with rat and human TARM-1 respectively.
  • TARM-1 associates with the ITAM bearing adaptor FcR gamma, but not with DAP10 or DAP12.
  • TARM1 expression is also up-regulated by bone marrow-derived macrophages and dendritic cells following stimulation with TLR agonists in vitro.
  • TARM1 receptor stimulation on macrophages and neutrophils co-stimulate the secretion of proinflammatory cytokines induced by TLR ligands, such as LPS.
  • TARM1 Fc fusion protein inhibits anti-CD3 induced CD4 + cell activation, suggesting that TARM1 ectodomain interacts with an unidentified receptor on T cells to inhibit T cell activation.

Specifications

Accession Number B6A8R8
For Use With (Application) Bioactivity
Gene ID (Entrez) 441864
Molecular Weight (g/mol) 53 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse OLT-2/TARM1 protein

Mouse TARM-1 (Gln17-Ile257) Accession # B6A8R8IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.