Learn More
R&D Systems™ Recombinant Mouse PDGF-D Protein
Description
Specifications
Specifications
| Accession Number | Q925I7.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl. |
| Gene ID (Entrez) | 80310 |
| Molecular Weight (g/mol) | 14 kDa |
| Name | IEGFMSTP036, Iris-expressed growth factor, PDGFD, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse PDGF-D protein Ser250-Arg370 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.