Learn More
Biomatik Corporation Recombinant Mouse Perforin-1 (Prf1)
Supplier:  Biomatik Corporation RPC25638
Encompass_Preferred
Recombinant Mouse Perforin-1 (Prf1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Prf1. Target Synonyms: CytolysinLymphocyte pore-forming proteinPFP. Accession Number: P10820. Expression Region: 21~554aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 63.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7086