Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Persephin Protein

Catalog No. p-4722014
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with Trehalose and with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl with Trehalose.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2479PS010CF Lyophilized from a 0.2μm filtered solution in HCl with Trehalose.
2479PS010 Lyophilized from a 0.2μm filtered solution in HCl with Trehalose and with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2479PS010CF Supplier R&D Systems™ Supplier No. 2479PS010/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Persephin Protein is derived from E. coli. The Recombinant Mouse Persephin Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl with Trehalose.
Gene ID (Entrez) 19197
Molecular Weight (g/mol) 10.3 kDa (monomer)
Name Persephin
Quantity 10 μg
Source E. coli-derived mouse Persephin protein Ala61-Gly156
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.