Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Proprotein Convertase 9/PCSK9 Protein

Catalog No. 9258SE020
Change view
Click to view available options
:
Recombinant Mouse Proprotein Convertase 9/PCSK9 Protein, Carrier-Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
92-58S-E020 Recombinant Mouse Proprotein Convertase 9/PCSK9 Protein, Carrier-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-58S-E020 Supplier R&D Systems™ Supplier No. 9258SE020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Mouse LDLR (Catalog # 2255-LD) is coated at 2 μg/mL, Recombinant Mouse Proprotein Convertase 9/PCSK9 (Catalog # 9258-SE) binds with an ED50 = 25-150 ng/mL.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number Q80W65
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 255738
Molecular Weight (g/mol) M.W. (Observed): 18 kDa (pro domain) and 57-67 kDa (mature form), reducing conditions; M.W. (theoretical): 14 kDa (pro domain) and 59 kDa (mature form)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived mouse Proprotein Convertase 9/PCSK9 protein Ser156-Gln694 with a C-terminal 10-His tag; Gln35-Gln155
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Proprotein Convertase 9/PCSK9
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.