Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse/Rat Noggin, Animal-Free Protein

Catalog No. QK0330500
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK0330500 500 μg
QK0330050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. QK0330500 Supplier R&D Systems™ Supplier No. QK0330500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
No significant difference between EC50 of reference and test lots

Specifications

Accession Number P97466.1
For Use With (Application) Bioactivity
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 9241
Molecular Weight (g/mol) M.W. (Observed): Oligomeric mouse Noggin protein only, M.W. (Theroretical): 46 kDa (dimer)
Quantity 500 μg
Source E. coli-derived Noggin protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired.
Protein Noggin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.