Learn More
Biomatik Corporation Recombinant Mouse Receptor-interacting serine/threonine-protein kinase 3 (Ripk3)
Supplier:  Biomatik Corporation RPC25552
Encompass_Preferred
Recombinant Mouse Receptor-interacting serine/threonine-protein kinase 3 (Ripk3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ripk3. Target Synonyms: RIP-like protein kinase 3Receptor-interacting protein 3; RIP-3; mRIP3. Accession Number: Q9QZL0. Expression Region: 1~486aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5312