Learn More
Medchemexpress LLC Recombinant mouse scavenger receptor class B member 1 (SR-BI/SCARB1) protein | >95.0% | 10 UG
Supplier: Medchemexpress LLC HYP7608610UG
Recombinant mouse SR-BI/SCARB1 extracellular domain expressed in HEK293 cells, supplied with C-terminal human Fc and His tags. Provided as a purified protein preparation for biochemical assays, ligand-binding studies, and antibody validation.
- Recombinant mouse SR-BI/SCARB1 protein with C-terminal human Fc and His tags.
- Expressed in HEK293 mammalian cells for proper folding and post-translational modifications.
- Purified to >95% as determined by reducing SDS-PAGE.
- Supplied as a 10 μg vial for small-scale assays and validation.
- Suitable for biochemical assays, ligand binding, and antibody validation.