Learn More
R&D Systems™ Recombinant Mouse Semaphorin 3C Fc Chimera (Truncated)
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris, Citric Acid, NaCl and Tween™ 20. |
| Gene ID (Entrez) | 20348 |
| Molecular Weight (g/mol) | 108.9 kDa (monomer) |
| Name | Semaphorin 3C Fc Chimera, Truncated |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Semaphorin 3C protein, (N-terminus) MHHHHHH Mouse Semaphorin 3C (Gln24-Ser741,Arg548Ala,Arg552Ala) Accession &Num; Q62181 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.