Learn More
R&D Systems™ Recombinant Mouse Semaphorin 4A mFc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q62178.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 64218 |
| Molecular Weight (g/mol) | M.W.-observed: 90-110 kDa, under reducing conditions., M.W.-theroretical: 99 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse Semaphorin 4 A protein Mouse Sema 4 A (Thr33-Phe683) (C-terminus) Accession # Q62178.2 IEGRMDP Mouse IgG2a (Glu98-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4 A, SEMA4A, SEMAB, semaphorin-4 A, Semaphorin-B, SemB, SEMBFLJ12287 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.