Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant mouse SIRP alpha (CD172a) Fc-fusion protein | >90.0% | 50 UG
SDP

Catalog No. 5000290882
Encompass_Preferred

Recombinant mouse SIRP alpha (CD172a) Fc-fusion protein produced in HEK293 cells, supplied as a purified Fc fusion for in vitro receptor-ligand interaction studies. The protein is provided as a 50 μg vial with reported purity greater than 90% and is suitable for biochemical and biophysical assays.

  • Recombinant ectodomain fused to Fc for improved stability and avidity.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • Purity greater than 90% for reliable experimental performance.
  • Compatible with ELISA, SPR/BLI, and other binding assays.
  • Useful for studying CD47-SIRPα interactions and immune signaling pathways.

Catalog No. 50-002-90882 Supplier Medchemexpress LLC Supplier No. HYP7607150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse SIRP alpha (CD172a) Fc-fusion protein produced in HEK293 cells, supplied as a purified Fc fusion for in vitro receptor-ligand interaction studies. The protein is provided as a 50 μg vial with reported purity greater than 90% and is suitable for biochemical and biophysical assays.

  • Recombinant ectodomain fused to Fc for improved stability and avidity.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • Purity greater than 90% for reliable experimental performance.
  • Compatible with ELISA, SPR/BLI, and other binding assays.
  • Useful for studying CD47-SIRPα interactions and immune signaling pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.