Learn More
R&D Systems™ Recombinant Mouse SIRP beta 1B Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | NP_001166931.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Molecular Weight (g/mol) | Observed: 70-95 kDa and 175-200 kDa, under reducing conditions; Theroretical: 64 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse SIRP beta 1 B protein Mouse SIRP beta 1 B (Met28-Gly360) Accession # NP_001166931.1(N-terminus) IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.