Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh Protein, Animal-Free

Catalog No. BTSHHAFL050
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTSHHAFL050 50 μg
BTSHHAFL500 500 μg
BTSHHAFL100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTSHHAFL050 Supplier R&D Systems™ Supplier No. BTSHHAFL050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Measured by its ability to induce alkaline phosphatase production by C3H10T1/2 mouse embryonic fibroblast cells. Nakamura, T. et al. (1997) Biochem. Biophys. Res. Commun. 237:465. The ED50 for this effect is 0.0500-0.600 μg/mL.

Specifications

Accession Number Q62226.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Sodium Phosphate, NaCl and DTT with Trehalose.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) Observed: 21 kDa, under reducing conditions.,Theoretical: 19.8 kDa
Quantity 50 μg
Source E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile) with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in water.
Protein Sonic Hedgehog/Shh
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.