Learn More
R&D Systems™ Recombinant Mouse Sphingosine Kinase 1/SPHK1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 20698 |
| Molecular Weight (g/mol) | 43 kDa |
| Name | Sphingosine Kinase 1/SPHK1 |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Sphingosine Kinase 1/SPHK1 protein Glu2-Pro382,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.