Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TEM8/ANTXR1 Fc Chimera Protein

Catalog No. 10202AR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-202-AR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-202-AR050 Supplier R&D Systems™ Supplier No. 10202AR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Anthrax toxin receptor 1 (ANTXR1), also known as Tumor endothelial marker 8 (TEM8), is a glycoprotein of the Anthrax Toxin Receptor family that is expressed by endothelial cells

  • TEM8/ANTXR1 has been implicated in tumor angiogenesis, as its expression has been shown to up-regulate in tumor blood vessels and is characterized as a tumor endothelial marker
  • TEM8/ANTXR1 was reported to be an amplifier of Wnt signaling in tumor microenvironment
  • Additionally, TEM8/ANTXR1 serves as the receptor for Seneca Valley virus, an oncolytic picornavirus affecting neuroendocrine cancers
  • Mouse TEM8/ANTXR1 shares 99% and 100% aa identity with human and rat TEM8/ANTXR1, respectively, within the extracellular domain

Specifications

Accession Number Q9CZ52
For Use With (Application) Bioactivity
Gene ID (Entrez) 84168
Molecular Weight (g/mol) 60 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse TEM8/ANTXR1 protein

Mouse TEM8/ANTXR1 (Glu31-Ser319) Accession # Q9CZ52IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.