Learn More
Biomatik Corporation Recombinant Mouse Triggering receptor expressed on myeloid cells 2 (Trem2), partial
Supplier:  Biomatik Corporation RPC23856
Encompass_Preferred
Recombinant Mouse Triggering receptor expressed on myeloid cells 2 (Trem2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Trem2. Target Synonyms: Triggering receptor expressed on monocytes 2. Accession Number: Q99NH8. Expression Region: 19~171aa. Tag Info: N-Terminal 6Xhis-Myc-Tagged. Theoretical MW: 20.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5019