Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Tryptase epsilon/BSSP-4 Protein

Catalog No. 2059SE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2059SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2059SE010 Supplier R&D Systems™ Supplier No. 2059SE010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a colorimetric peptide substrate, N-carbobenzyloxy-Arg-ThioBenzyl ester (Z-R-SBzl), in the presence of 5,5’Dithio-bis (2-nitrobenzoic acid) (DTNB). Edwards, K.M. et al. (1999) J. Biol. Chem. 274:30468. The specific activity is >5,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q7TML0
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in MES and NaCl.
Gene ID (Entrez) 64063
Molecular Weight (g/mol) M.W. (Observed): 37 kDa and 33 kDa, reducing conditions; M.W. (theoretical): 32 kDa
Name Tryptase epsilon/BSSP-4
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived mouse Tryptase epsilon/BSSP-4 protein Ala33-Ser306, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Pro form
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Prss22
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile 50 mM Tris, 10 mM CaCl2 and 150 mM NaCl, pH 7.5.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Tryptase epsilon/BSSP-4/PRSS22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.