Learn More
Creative Biomart GMP Recombinant Mouse VEGFA protein, His-tag
Supplier:  Creative Biomart VEGFA134THP
Encompass_Preferred
Recombinant Mouse VEGFA protein GMP Grade(NP_001273986.1)(170 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5278