Learn More
R&D Systems™ Recombinant Mouse VEGFR3/Flt-4 Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 14257 |
| Molecular Weight (g/mol) | Predicted MW: 112kDa (monomer) Observed MW: 123kDa with two additional minor bands at 73kDa and 60kDa, reducing conditions |
| Name | VEGFR3/Flt-4 |
| Quantity | 100μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse VEGFR3/Flt-4 protein, (N-terminus) Met Mouse VEGFR3 (Tyr25-Asp770) Accession &Num; P35917 IEGRDID Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, FLT4, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.