Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse VSIG4 Fc Chimera Protein

Catalog No. 4674VS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4674VS050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4674VS050 Supplier R&D Systems™ Supplier No. 4674VS050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit anti-CD3-induced proliferation of splenic T lymphocytes. Vogt, L. et al. (2006) J. Clin. Invest. 116:2817. The presence of immobilized Recombinant Mouse VSIG4 Fc Chimera inhibited the anti-CD3 response by =50%. The ED50 for this effect is 1.25-5 μg/mL.

Specifications

Accession Number NP_808457
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11326
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 46 kDa (monomer)
Name VSIG4 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse VSIG4 protein Mouse VSIG4 (His20-Pro187) (N-terminus) Accession # NP_808457 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Vsig4
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VSIG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.