Learn More
R&D Systems™ Recombinant Mouse VSIG8 Fc Chimera Protein
Description
Order Info
Ships at ambient temperature
Specifications
Specifications
| Accession Number | Q6P3A4 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 391123 |
| Molecular Weight (g/mol) | M.W. (Observed): 56-66 kDa, reducing conditions; M.W. (theoretical): 54 kDa |
| Name | VSIG8 Fc Chimera Recombinant Protein |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse VSIG8 protein Mouse VSIG8 (Val22-Gly262) (N-terminus) Accession # Q6P3A4 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.