Learn More
R&D Systems™ Recombinant Mouse Wnt-3/sFRP-1 Complex Protein
Description
Specifications
Specifications
| Accession Number | P17553.1, Q8C4U3.3 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS and NaCl with Trehalose. |
| Gene ID (Entrez) | 7473 |
| Molecular Weight (g/mol) | M.W.-observed: 36-45 kDa, under reducing conditions., M.W.-theroretical: 37 kDa (Wnt-3) & 33 kDa (sFRP-1) |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse Wnt-3 protein Mouse Wnt-3 (Gly22-Lys355) (C-terminus) Accession # P17553.1 Mouse sFRP-1 (Ser32-Lys314) (C-terminus) Accession # Q8C4U3.3 (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | FrzA, INT4, INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, SARP-2, sFRP1, TETAMS, wingless-type MMTV integration site family, member 3, WNT3, WNT-3 proto-oncogene protein |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.