Learn More
Active Motif Recombinant NFκB1 p50 (1-434) protein
SDP
Supplier:  Active Motif 81032
Encompass
Recombinant NFkB1 / p50 (1-434) protein including amino acid 1 - 434 of NFkB1 p105 subunit (accession number NP_003989.2) was expressed in E. coli cells with an N-terminal GST tag. The molecular weight of NFkB1 / p50 (1-434) is 73.9 kDa.
Catalog No. NC1851029