Learn More
Creative Biomart Anakinra, Recombinant Human IL1Ra
Catalog No. 502084161
Encompass_Preferred
The product is a recombinant, nonglycosylated human interleukin-1 receptor antagonist (IL-1Ra). The difference between the product and the native human IL-1Ra is that the product has an extra methionine residue at the amino terminus. It is manufactured by using the E. coli expression system. The product is composed of 153 amino acid residues.
Catalog No. 50-208-4161 Supplier Creative Biomart Supplier No. THP0110
Add to Cart
Add to Cart
The product is a recombinant, nonglycosylated human interleukin-1 receptor antagonist (IL-1Ra). The difference between the product and the native human IL-1Ra is that the product has an extra methionine residue at the amino terminus. It is manufactured by using the E. coli expression system. The product is composed of 153 amino acid residues.