Learn More
Abcam Recombinant Norovirus GII.4VP1 virus-like particles, 100UG
Supplier:  Abcam AB256447100UG
Encompass_Preferred
Recombinant Norovirus GII.4VP1 virus-like particles is a Full Length protein, expressed in HEK 293, with >95% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: VP1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1977752