Learn More
Active Motif Recombinant p300 protein
SDP
Supplier:  Active Motif 81158
Encompass
Recombinant p300 protein was expressed in baculovirus expression system as full-length protein (accession number NP_001420.2) with an N-terminal FLAG tag. The molecular weight of the protein is 265.4 kDa.
Catalog No. NC1851869