Learn More
Active Motif Recombinant p53 (TP53) protein
SDP
Supplier:  Active Motif 81091
Encompass
Recombinant p53 (TP53) protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_000537.3) with an N-terminal FLAG tag. The molecular weight of the protein is 44.9 kDa.
Catalog No. NC1851463