Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant PDGF-BB protein, cynomolgus (HEK293, N-mFc) | 98.0% | 10 UG
SDP

Catalog No. 5000291148
Encompass_Preferred

Recombinant PDGF-BB protein from cynomolgus sequence, produced in HEK293 cells and supplied as an N-terminal mouse Fc fusion. Supplied lyophilized from a PBS (pH 7.4) solution, it is characterized for purity, molecular weight, endotoxin level, and biological activity for use in cell signaling and proliferation research.

  • Expressed in HEK293 cells as N-terminal mFc fusion
  • Supplied lyophilized from PBS, pH 7.4
  • Purity 98.0% as determined by reducing SDS-PAGE
  • Molecular weight approximately 45-50 kDa (glycosylated)
  • Endotoxin <1 EU/μg (LAL method)
  • Biological activity ED50 5.317 ng/mL (cell proliferation assay)
  • Recommended storage lyophilized at -20°C; avoid repeated freeze-thaw cycles
  • Shipped on dry ice

Catalog No. 50-002-91148 Supplier Medchemexpress LLC Supplier No. HYP75966A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant PDGF-BB protein from cynomolgus sequence, produced in HEK293 cells and supplied as an N-terminal mouse Fc fusion. Supplied lyophilized from a PBS (pH 7.4) solution, it is characterized for purity, molecular weight, endotoxin level, and biological activity for use in cell signaling and proliferation research.

  • Expressed in HEK293 cells as N-terminal mFc fusion
  • Supplied lyophilized from PBS, pH 7.4
  • Purity 98.0% as determined by reducing SDS-PAGE
  • Molecular weight approximately 45-50 kDa (glycosylated)
  • Endotoxin <1 EU/μg (LAL method)
  • Biological activity ED50 5.317 ng/mL (cell proliferation assay)
  • Recommended storage lyophilized at -20°C; avoid repeated freeze-thaw cycles
  • Shipped on dry ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.