Learn More
Active Motif Recombinant PHD2 (EGLN1) protein
SDP
Supplier:  Active Motif 81065
Encompass
Recombinant human PHD2 (EGLN1) protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_071334.1) with an N-terminal FLAG tag. The molecular weight of the protein is 47 kDa.
Catalog No. NC1482261