Learn More
Active Motif RECOMBINANT PRC2 COMPLEX,20UG
SDP
Catalog No. NC1930921
Encompass
Recombinant PRC2 complex that includes full-length EZH2, SUZ12, EED and RbAp46/48 (accession numbers NP_001190176.1, NP_056170, NP_003788.2, NP_002884.1, and NP_005601.1, respectively) was expressed in Sf9 and contains an N-terminal FLAG tag at the N-terminus of EZH2. The molecular weights of expressed EZH2, SUZ12, EED and RbAp46/48 are 87 kDa, 83 kDa, 50.2 kDa, 47.8 kDa and 47.7 kDa, respectively. PRC2 (Polycomb Repressive Complex 2) is one of the two classes Polycomb-group, or PcG proteins (the other being PRC1) that are important epigenetic determinants of stem cell identity. They play an important role in long-term epigenetic silencing of genes during cell fate determination and differentiation. PRC2 functions as a repressor of chromatin. PRC2 is required to target recruitment to specific DNA sequences (termed Polycomb Response Elements or PREs) of genomic regions to be silenced.
Catalog No. NC1930921 Supplier Active Motif Supplier No. 31387
Add to Cart
Add to Cart
Recombinant PRC2 complex that includes full-length EZH2, SUZ12, EED and RbAp46/48 (accession numbers NP_001190176.1, NP_056170, NP_003788.2, NP_002884.1, and NP_005601.1, respectively) was expressed in Sf9 and contains an N-terminal FLAG tag at the N-terminus of EZH2. The molecular weights of expressed EZH2, SUZ12, EED and RbAp46/48 are 87 kDa, 83 kDa, 50.2 kDa, 47.8 kDa and 47.7 kDa, respectively. PRC2 (Polycomb Repressive Complex 2) is one of the two classes Polycomb-group, or PcG proteins (the other being PRC1) that are important epigenetic determinants of stem cell identity. They play an important role in long-term epigenetic silencing of genes during cell fate determination and differentiation. PRC2 functions as a repressor of chromatin. PRC2 is required to target recruitment to specific DNA sequences (termed Polycomb Response Elements or PREs) of genomic regions to be silenced.