Learn More
ACROBiosystems Human CD39 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems CD9H5253100UG
Encompass_Preferred
Recombinant Protein;100;Human CD39, Mouse IgG2a Fc Tag (CD9-H5253) is expressed from human 293 cells (HEK293). It contains AA Thr 38 - Val 478 (Accession # P49961-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 77.2 kDa.
Catalog No. 50-201-9490