Learn More
ACROBiosystems Cynomolgus Mesothelin / MSLN (296-580) Protein, His Tag (MALS verified)
Catalog No. 502019686
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Mesothelin (296-580), His Tag (MSN-C52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 296 - Gly 580 (Accession # A0A2K5TW94-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Mesothelin (296-580), His Tag (Cat. No. MSN-C52H6) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human MSLN Antibody, Human IgG1 with a linear range of 0.06-2 ng/mL (QC tested).;Immobilized Cynomolgus Mesothelin (296-580), His Tag (Cat. No. MSN-C52H6) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CA125, Fc,Avitag (Cat. No. CA5-H82F4) with a linear range of 2-39 ng/mL (Routinely tested).
Catalog No. 50-201-9686 Supplier ACROBiosystems Supplier No. MSNC52H6100UG
Recombinant Protein;100UG;Cynomolgus Mesothelin (296-580), His Tag (MSN-C52H6) is expressed from human 293 cells (HEK293). It contains AA Asp 296 - Gly 580 (Accession # A0A2K5TW94-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus Mesothelin (296-580), His Tag (Cat. No. MSN-C52H6) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human MSLN Antibody, Human IgG1 with a linear range of 0.06-2 ng/mL (QC tested).;Immobilized Cynomolgus Mesothelin (296-580), His Tag (Cat. No. MSN-C52H6) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CA125, Fc,Avitag (Cat. No. CA5-H82F4) with a linear range of 2-39 ng/mL (Routinely tested).