Learn More
ACROBiosystems Cynomolgus IL-17F / IL-24 Protein, His Tag
Supplier:  ACROBiosystems ILFC5243100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus IL-17F, His Tag (ILF-C5243) is expressed from human 293 cells (HEK293). It contains AA Arg 31 - Gln 163 (Accession # G7P4V0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus IL-17F, His Tag (Cat. No. ILF-C5243) at 2 μg/mL (100 μL/well) can bind Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) with a linear range of 0.005-0.313 μg/mL (QC tested).;Immobilized Cynomolgus IL-17F, His Tag (Cat. No. ILF-C5243) at 2 μg/mL (100 μL/well) can bind Recombinant Human IL-17RC Isoform 1 Fc Chimera with a linear range of 0.5-16 ng/mL (Routinely tested).
Catalog No. 50-201-9779