Learn More
ACROBiosystems Cynomolgus CD73 Protein, His Tag
Catalog No. 502019485
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus CD73, His Tag (CD3-C52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Cys 554 (Accession # EHH53214.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa.
Catalog No. 50-201-9485 Supplier ACROBiosystems Supplier No. CD3C52H9100UG
Recombinant Protein;100UG;Cynomolgus CD73, His Tag (CD3-C52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Cys 554 (Accession # EHH53214.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.3 kDa.