Learn More
ACROBiosystems Cynomolgus IL-17A / CTLA-8 Protein, His Tag
Supplier:  ACROBiosystems ILAC5249100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus IL-17A, His Tag (ILA-C5249) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (Accession # G7P4U9-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 18-19 kDa,24-25 kDa and 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9554