Learn More
ACROBiosystems Cynomolgus NKG2C / CD159c Protein, His Tag
Supplier:  ACROBiosystems NKCC5247100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus NKG2C, His Tag (NKC-C5247) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (Accession # Q68VD0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa.
Catalog No. 50-201-9437