Learn More
ACROBiosystems HCoV-229E S1 protein, His Tag
Catalog No. 502019594
Encompass_Preferred
Recombinant Protein;100UG;HCoV-229E S1 protein, His Tag (SIN-V52H4) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Ala 536 (Accession # P15423-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.8 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-201-9594 Supplier ACROBiosystems Supplier No. SINV52H4100UG
Recombinant Protein;100UG;HCoV-229E S1 protein, His Tag (SIN-V52H4) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Ala 536 (Accession # P15423-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.8 kDa. The protein migrates as 70-110 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.