Learn More
ACROBiosystems Human NKG2C&CD94 Protein, Fc Tag
Supplier:  ACROBiosystems NC4H5258100UG
Encompass_Preferred
Recombinant Protein;100UG;Human NKG2C&CD94, Fc Tag (NC4-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (NKG2C) & Asp 57 - Ile 179 (CD94) (Accession # P26717-1 (NKG2C) & Q13241-1 (CD94)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NKG2C&CD94, Fc Tag (Cat. No. NC4-H5258) at 5 μg/mL (100 μL/well) can bind HLA-E*01:03 HLA-A leader3-11 Tetramer-VMAPRTLVL-PE,TS-ME01-1 (QC tested).
Catalog No. 50-201-9549