Learn More
ACROBiosystems Human IFN-alpha 1 Protein, His Tag
Supplier:  ACROBiosystems IFAH52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IFN-alpha 1, His Tag (IFA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 189 (Accession # P01562-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.2 kDa.
Catalog No. 50-201-9564