Learn More
ACROBiosystems Human Siglec-1 / CD169 Protein, His Tag
Catalog No. 502019572
Encompass_Preferred
Recombinant Protein;100UG;Human Siglec-1, His Tag (SG1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Gln 1641 (Accession # Q9BZZ2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 182.5 kDa.
Catalog No. 50-201-9572 Supplier ACROBiosystems Supplier No. SG1H52H3100UG
Recombinant Protein;100UG;Human Siglec-1, His Tag (SG1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Gln 1641 (Accession # Q9BZZ2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 182.5 kDa.